siRNA Details
| Gene Symbol: NUDT15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018283.3 | NP_060753.1 | 3 | 726 | ||
| GenBank mRNA | AK001818.1 | - | 3 | 726 | ||
| BC064607.1 | - | 3 | 545 | |||
| BC107875.1 | - | 3 | 599 | 6451308 | ||
| BC133015.1 | - | 3 | 667 | 40147037 | ||
| BC133017.1 | - | 3 | 667 | 40147039 | ||

NUDT15
Genomic Map