siRNA Details
| Gene Symbol: LRRN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099658.1 | NP_001093128.1 | 3 | 1038 | ||
| NM_001099660.1 | NP_001093130.1 | 4 | 1145 | |||
| NM_018334.4 | NP_060804.3 | 2 | 956 | |||
| GenBank mRNA | AB060967.1 | - | 1 | 405 | ||
| AF134481.1 | - | 1 | 436 | |||
| AK001991.1 | - | 4 | 1057 | |||
| AK292556.1 | - | 2 | 545 | |||
| AL442092.1 | - | 1 | 361 | |||
| AY358280.1 | - | 3 | 578 | |||
| BC035133.2 | - | 2 | 835 | 5263900 | ||
| CR457397.1 | - | 1 | 99 | |||

LRRN3
Genomic Map