siRNA Details
| Gene Symbol: FBXL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018378.2 | NP_060848.2 | 3 | 347 | ||
| GenBank mRNA | AK002140.1 | - | 3 | 347 | ||
| BC014414.1 | - | 3 | 347 | 4548146 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FBXL8| Gene Symbol: FBXL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018378.2 | NP_060848.2 | 3 | 347 | ||
| GenBank mRNA | AK002140.1 | - | 3 | 347 | ||
| BC014414.1 | - | 3 | 347 | 4548146 | ||