siRNA Details
| Gene Symbol: IL26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018402.1 | NP_060872.1 | 1, 2 | 203 | ||
| GenBank mRNA | AJ251549.1 | - | 1, 2 | 203 | ||
| BC066270.1 | - | 1, 2 | 202 | 7002121 | ||
| BC066271.1 | - | 1, 2 | 202 | 7002122 | ||

IL26
Genomic Map