siRNA Details
| Gene Symbol: EAPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318916.1 | NP_001305845.1 | 5 | 847 | ||
| NM_018453.3 | NP_060923.2 | 6 | 965 | |||
| XM_011536958.1 | XP_011535260.1 | 6 | 1243 | |||
| GenBank mRNA | AF217512.1 | - | 6 | 955 | ||
| AK000585.1 | - | 6 | 929 | |||
| AK225467.1 | - | 6 | 920 | |||
| AK225477.1 | - | 6 | 929 | |||
| AK225991.1 | - | 6 | 929 | |||
| AY157300.1 | - | 6 | 951 | |||
| AY157301.1 | - | 6 | 951 | |||
| AY869694.1 | - | 6 | 888 | |||
| BC001245.1 | - | 6 | 938 | 3460193 | ||

EAPP
Genomic Map