siRNA Details
| Gene Symbol: FOXN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085471.1 | NP_001078940.1 | 7 | 6652 | ||
| NM_005197.3 | NP_005188.2 | 6 | 6703 | |||
| GenBank mRNA | AF116643.2 | - | 1 | 577 | ||
| AK123929.1 | - | 1 | 1328 | |||
| AK314098.1 | - | 1 | 638 | |||
| BC010460.1 | - | 1 | 443 | |||

FOXN3
Genomic Map