siRNA Details
| Gene Symbol: WSB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278557.1 | NP_001265486.1 | 3 | 280 | ||
| NM_018639.4 | NP_061109.1 | 3 | 380 | |||
| XM_017019642.1 | XP_016875131.1 | 3 | 282 | |||
| GenBank mRNA | AF163324.1 | - | 2 | 217 | ||
| AF229181.1 | - | 3 | 319 | |||
| AK294230.1 | - | 2 | 138 | |||
| AK295555.1 | - | 3 | 281 | |||
| AK310892.1 | - | 3 | 258 | |||
| BC015887.2 | - | 3 | 270 | 3910968 | ||

WSB2
Genomic Map