siRNA Details
| Gene Symbol: WSB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278557.1 | NP_001265486.1 | 4, 5 | 617 | ||
| NM_018639.4 | NP_061109.1 | 4, 5 | 717 | |||
| XM_017019642.1 | XP_016875131.1 | 4, 5 | 619 | |||
| GenBank mRNA | AF163324.1 | - | 3, 4 | 554 | ||
| AF229181.1 | - | 4, 5 | 656 | |||
| AK295555.1 | - | 4, 5 | 618 | |||
| AK310892.1 | - | 4, 5 | 595 | |||
| BC015887.2 | - | 4, 5 | 607 | 3910968 | ||

WSB2
Genomic Map