siRNA Details
| Gene Symbol: PCDHGB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018923.2 | NP_061746.1 | 1 | 1312 | ||
| NM_032096.1 | NP_115267.1 | 1 | 1312 | |||
| GenBank mRNA | AF152331.1 | - | 1 | 1312 | ||
| AF152518.1 | - | 1 | 1312 | |||
| BC101805.1 | - | 1 | 1375 | 8069311 | ||

PCDHGB2
Genomic Map