siRNA Details
| Gene Symbol: HIST1H3D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003530.4 | NP_003521.2 | 2 | 866 | ||
| GenBank mRNA | BC031333.1 | - | 2 | 785 | 3608479 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST1H3D| Gene Symbol: HIST1H3D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003530.4 | NP_003521.2 | 2 | 866 | ||
| GenBank mRNA | BC031333.1 | - | 2 | 785 | 3608479 | |