siRNA Details
| Gene Symbol: PCDHB15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018935.3 | NP_061758.1 | 1 | 2844 | ||
| GenBank mRNA | AF217742.1 | - | 1 | 2614 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCDHB15| Gene Symbol: PCDHB15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018935.3 | NP_061758.1 | 1 | 2844 | ||
| GenBank mRNA | AF217742.1 | - | 1 | 2614 | ||