siRNA Details
| Gene Symbol: PCDHB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018937.4 | NP_061760.1 | 1 | 1392 | ||
| GenBank mRNA | AF152496.1 | - | 1 | 984 | ||
| AF217755.1 | - | 1 | 984 | |||
| AK313451.1 | - | 1 | 1161 | |||
| BC104817.1 | - | 1 | 1011 | 8143820 | ||

PCDHB3
Genomic Map