siRNA Details
| Gene Symbol: PCDHB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018937.4 | NP_061760.1 | 1 | 3316 | ||
| GenBank mRNA | AF217755.1 | - | 1 | 2908 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCDHB3| Gene Symbol: PCDHB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018937.4 | NP_061760.1 | 1 | 3316 | ||
| GenBank mRNA | AF217755.1 | - | 1 | 2908 | ||