siRNA Details
| Gene Symbol: PCDHB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018938.3 | NP_061761.1 | 1 | 341 | ||
| GenBank mRNA | AF152497.1 | - | 1 | 92 | ||
| AF217754.1 | - | 1 | 92 | |||
| AK313640.1 | - | 1 | 251 | |||
| BC098139.1 | - | 1 | 92 | 40008131 | ||
| BC098346.1 | - | 1 | 92 | 40008130 | ||
| DA231000.1 | - | 1 | 342 | |||
| DA409037.1 | - | 1 | 280 | |||

PCDHB4
Genomic Map