siRNA Details
| Gene Symbol: TAS2R5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018980.2 | NP_061853.1 | 1 | 428 | ||
| GenBank mRNA | BC069509.1 | - | 1 | 283 | 7262216 | |
| BC095522.1 | - | 1 | 428 | 30706589 | ||
| BC101586.1 | - | 1 | 304 | 8069092 | ||
| BC101588.1 | - | 1 | 304 | 8069094 | ||

TAS2R5
Genomic Map