siRNA Details
| Gene Symbol: MAGEL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019066.4 | NP_061939.3 | 1 | 3364 | ||
| GenBank mRNA | AJ243531.1 | - | 1 | 1349 | ||
| BC035839.1 | - | 1 | 1486 | |||
| BC063834.1 | - | 1 | 1546 | |||
| BC112257.1 | - | 1 | 1286 | |||
| BC112259.1 | - | 1 | 1286 | |||

MAGEL2
Genomic Map