siRNA Details
| Gene Symbol: SPATA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286238.1 | NP_001273167.1 | 8 | 1008 | ||
| NM_001286239.1 | NP_001273168.1 | 7 | 966 | |||
| NM_019073.3 | NP_061946.1 | 8 | 1008 | |||
| XM_006710699.2 | XP_006710762.1 | 8 | 1002 | |||
| XM_006710700.2 | XP_006710763.1 | 6 | 807 | |||
| XM_006710701.3 | XP_006710764.1 | 8 | 1002 | |||
| XM_011541606.1 | XP_011539908.1 | 8 | 1002 | |||
| XM_011541607.2 | XP_011539909.1 | 5 | 571 | |||
| XM_011541608.1 | XP_011539910.1 | 7 | 945 | |||
| XM_011541609.2 | XP_011539911.1 | 6 | 653 | |||
| XM_017001509.1 | XP_016856998.1 | 8 | 1002 | |||
| XM_017001510.1 | XP_016856999.1 | 5 | 757 | |||
| XM_017001511.1 | XP_016857000.1 | 8 | 1002 | |||
| XM_017001512.1 | XP_016857001.1 | 9 | 851 | |||
| XM_017001513.1 | XP_016857002.1 | 6 | 615 | |||
| XM_017001514.1 | XP_016857003.1 | 9 | 893 | |||
| XM_017001515.1 | XP_016857004.1 | 8 | 830 | |||
| GenBank mRNA | AK000869.1 | - | 8 | 973 | ||
| AK301770.1 | - | 7 | 909 | |||
| BC020660.1 | - | 8 | 810 | 4657425 | ||

SPATA6
Genomic Map