siRNA Details
| Gene Symbol: KIAA1217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019590.4 | NP_062536.2 | 19 | 4344 | ||
| XM_005252516.3 | XP_005252573.1 | 14 | 3250 | |||
| XM_011519552.2 | XP_011517854.2 | 20 | 4399 | |||
| XM_011519555.2 | XP_011517857.2 | 19 | 4317 | |||
| XM_011519558.2 | XP_011517860.2 | 19 | 4281 | |||
| XM_011519559.2 | XP_011517861.2 | 18 | 3986 | |||
| XM_017016416.1 | XP_016871905.1 | 20 | 4425 | |||
| XM_017016417.1 | XP_016871906.1 | 18 | 4239 | |||
| XM_017016418.1 | XP_016871907.1 | 18 | 5061 | |||
| XM_017016419.1 | XP_016871908.1 | 17 | 4956 | |||
| XM_017016420.1 | XP_016871909.1 | 18 | 3808 | |||
| XM_017016421.1 | XP_016871910.1 | 20 | 4018 | |||
| XM_017016422.1 | XP_016871911.1 | 17 | 3703 | |||
| XM_017016423.1 | XP_016871912.1 | 15 | 3355 | |||
| XM_017016424.1 | XP_016871913.1 | 15 | 3427 | |||
| GenBank mRNA | AK125675.1 | - | 1 | 201 | ||
| AK300422.1 | - | 14 | 3261 | |||
| AL833280.1 | - | 19 | 3972 | |||
| BX640796.1 | - | 14 | 3251 | |||
| DB164876.1 | - | 1 | 103 | |||
 Back To Top
 Back To Top
 KIAA1217
KIAA1217 Genomic Map
Genomic Map Back To Top
 Back To Top