siRNA Details
| Gene Symbol: CPXM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184699.1 | NP_001171628.1 | 11 | 1518 | ||
| NM_019609.4 | NP_062555.1 | 11 | 1518 | |||
| GenBank mRNA | AK027661.1 | - | 11 | 1454 | ||
| AK075508.1 | - | 7 | 730 | |||
| AK303142.1 | - | 8 | 997 | |||
| AK315027.1 | - | 11 | 1505 | |||
| AY358956.1 | - | 11 | 1474 | |||
| BC063430.1 | - | 11 | 1495 | 5239864 | ||

CPXM1
Genomic Map