siRNA Details
Gene Symbol: CCL28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301873.1 | NP_001288802.1 | 4 | 1165 | ||
NM_001301874.1 | NP_001288803.1 | 4 | 569 | |||
GenBank mRNA | AF110384.1 | - | 4 | 1144 | ||
AF266504.1 | - | 4 | 546 |
