siRNA Details
| Gene Symbol: FSTL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128427.2 | NP_001121899.1 | 5 | 954 | ||
| NM_001128428.2 | NP_001121900.1 | 5 | 954 | |||
| NM_020116.4 | NP_064501.2 | 5 | 957 | |||
| XM_011532126.1 | XP_011530428.1 | 5 | 923 | |||
| GenBank mRNA | AB033089.1 | - | 5 | 884 | ||
| AK315152.1 | - | 5 | 925 | |||
| AL137695.1 | - | 2 | 301 | |||
| BC036502.2 | - | 5 | 958 | 5265170 | ||
| BC073978.1 | - | 5 | 658 | |||

FSTL5
Genomic Map