siRNA Details
| Gene Symbol: KCMF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020122.4 | NP_064507.3 | 7 | 1966 | ||
| XM_006712052.3 | XP_006712115.1 | 7 | 1998 | |||
| XM_011532990.1 | XP_011531292.1 | 7 | 1888 | |||
| XM_017004511.1 | XP_016860000.1 | 6 | 1602 | |||
| GenBank mRNA | AF131740.1 | - | 1 | 221 | ||
| AK023403.1 | - | 7 | 1645 | |||
| AK311099.1 | - | 1 | 507 | |||

KCMF1
Genomic Map