siRNA Details
| Gene Symbol: IFNK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020124.2 | NP_064509.2 | 2 | 823 | ||
| GenBank mRNA | AF146759.1 | - | 2 | 845 | ||
| AF315688.1 | - | 2 | 834 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IFNK| Gene Symbol: IFNK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020124.2 | NP_064509.2 | 2 | 823 | ||
| GenBank mRNA | AF146759.1 | - | 2 | 845 | ||
| AF315688.1 | - | 2 | 834 | |||