siRNA Details
| Gene Symbol: THAP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020147.3 | NP_064532.1 | 1, 2 | 590 | ||
| GenBank mRNA | AK313472.1 | - | 1, 2 | 588 | ||
| AL360202.1 | - | 1, 2 | 549 | |||
| BC027857.1 | - | 1, 2 | 571 | 5222191 | ||
| BC072414.1 | - | 1, 2 | 578 | 6167441 | ||
| DA272000.1 | - | 1, 2 | 590 | |||

THAP10
Genomic Map