siRNA Details
| Gene Symbol: THAP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020147.3 | NP_064532.1 | 3 | 1853 | ||
| GenBank mRNA | AL360202.1 | - | 3 | 1812 | ||
| BC027857.1 | - | 3 | 1834 | 5222191 | ||
| BC072414.1 | - | 3 | 1841 | 6167441 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

THAP10| Gene Symbol: THAP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020147.3 | NP_064532.1 | 3 | 1853 | ||
| GenBank mRNA | AL360202.1 | - | 3 | 1812 | ||
| BC027857.1 | - | 3 | 1834 | 5222191 | ||
| BC072414.1 | - | 3 | 1841 | 6167441 | ||