siRNA Details
| Gene Symbol: CCDC177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271507.1 | NP_001258436.1 | 2 | 3655 | ||
| GenBank mRNA | AF284217.1 | - | 1 | 864 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC177| Gene Symbol: CCDC177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271507.1 | NP_001258436.1 | 2 | 3655 | ||
| GenBank mRNA | AF284217.1 | - | 1 | 864 | ||