siRNA Details
| Gene Symbol: SDHAF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020186.2 | NP_064571.1 | 2 | 1794 | ||
| GenBank mRNA | AF201933.1 | - | 2 | 683 | ||
| BC020621.1 | - | 2 | 676 | 4766494 | ||
| BC028409.1 | - | 2 | 1802 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SDHAF3| Gene Symbol: SDHAF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020186.2 | NP_064571.1 | 2 | 1794 | ||
| GenBank mRNA | AF201933.1 | - | 2 | 683 | ||
| BC020621.1 | - | 2 | 676 | 4766494 | ||
| BC028409.1 | - | 2 | 1802 | |||