siRNA Details
| Gene Symbol: MRPS22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020191.2 | NP_064576.1 | 1 | 131 | ||
| XM_005247640.2 | XP_005247697.1 | 1 | 130 | |||
| XM_006713703.3 | XP_006713766.1 | 1 | 156 | |||
| GenBank mRNA | AB208981.1 | - | 1 | 49 | ||
| AF063603.1 | - | 1 | 128 | |||
| AF226045.1 | - | 1 | 131 | |||
| AF321613.1 | - | 3 | 555 | |||
| AK075436.1 | - | 1 | 123 | |||
| AK292852.1 | - | 1 | 130 | |||
| BC009296.2 | - | 1 | 126 | 4123476 | ||

MRPS22
Genomic Map