siRNA Details
| Gene Symbol: NT5M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020201.3 | NP_064586.1 | 5 | 1405 | ||
| XM_005256731.3 | XP_005256788.1 | 6 | 1469 | |||
| XM_005256732.3 | XP_005256789.1 | 5 | 1412 | |||
| XM_005256737.1 | XP_005256794.1 | 6 | 1225 | |||
| XM_011523963.1 | XP_011522265.1 | 7 | 1262 | |||
| XM_011523964.2 | XP_011522266.1 | 7 | 1231 | |||
| XM_011523965.2 | XP_011522267.1 | 6 | 1135 | |||
| XM_011523967.2 | XP_011522269.1 | 6 | 1505 | |||
| XM_017024879.1 | XP_016880368.1 | 6 | 2165 | |||
| GenBank mRNA | AF210652.1 | - | 5 | 1436 | ||
| BC035838.1 | - | 5 | 1379 | 5727827 | ||

NT5M
Genomic Map