siRNA Details
| Gene Symbol: CASS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164114.1 | NP_001157586.1 | 5 | 1261 | ||
| NM_001164116.1 | NP_001157588.1 | 6 | 1447 | |||
| NM_020356.3 | NP_065089.2 | 5 | 1423 | |||
| XM_006723831.3 | XP_006723894.1 | 6 | 1847 | |||
| GenBank mRNA | AJ276678.1 | - | 6 | 1444 | ||
| AK295613.1 | - | 5 | 1126 | |||
| AK308057.1 | - | 5 | 1424 | |||
| AK314653.1 | - | 5 | 1356 | |||
| BC027951.1 | - | 5 | 1373 | 5205865 | ||

CASS4
Genomic Map