siRNA Details
| Gene Symbol: GPR84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020370.2 | NP_065103.1 | 2 | 1002 | ||
| XM_011538495.2 | XP_011536797.1 | 2 | 1689 | |||
| GenBank mRNA | AF237762.1 | - | 2 | 968 | ||
| AF282693.1 | - | 2 | 990 | |||
| AK290394.1 | - | 1 | 1536 | |||
| BC020614.1 | - | 2 | 1000 | 4279185 | ||
| FJ348261.1 | - | 1 | 900 | |||
More Information
Set Membership: |
|

GPR84
Genomic Map