siRNA Details
| Gene Symbol: AVEN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020371.2 | NP_065104.1 | 6 | 1088 | ||
| XM_011521818.2 | XP_011520120.1 | 7 | 1271 | |||
| XM_011521819.2 | XP_011520121.1 | 6 | 1314 | |||
| XM_011521820.1 | XP_011520122.1 | 6 | 789 | |||
| GenBank mRNA | AF283508.1 | - | 6 | 1084 | ||
| AK025888.1 | - | 1 | 51 | |||
| BC010488.1 | - | 6 | 1082 | 2900586 | ||
| BC063533.1 | - | 6 | 1072 | 4425400 | ||

AVEN
Genomic Map