siRNA Details
| Gene Symbol: CYSLTR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308465.1 | NP_001295394.1 | 6 | 2012 | ||
| NM_001308467.1 | NP_001295396.1 | 6 | 1989 | |||
| NM_001308468.1 | NP_001295397.1 | 4 | 1810 | |||
| NM_001308469.1 | NP_001295398.1 | 4 | 1787 | |||
| NM_001308470.1 | NP_001295399.1 | 3 | 1720 | |||
| NM_001308471.1 | NP_001295400.1 | 2 | 1619 | |||
| NM_001308476.1 | NP_001295405.1 | 5 | 1883 | |||
| NM_020377.3 | NP_065110.1 | 5 | 1860 | |||
| GenBank mRNA | AB038269.1 | - | 1 | 1574 | ||
| AB041947.1 | - | 1 | 1645 | |||
| AK291739.1 | - | 5 | 1860 | |||
| BC069160.1 | - | 1 | 1314 | 7216884 | ||
More Information
Set Membership: |
|

CYSLTR2
Genomic Map