siRNA Details
| Gene Symbol: PDSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020381.3 | NP_065114.3 | 8 | 1919 | ||
| XM_011535956.2 | XP_011534258.1 | 9 | 2215 | |||
| XM_011535957.2 | XP_011534259.1 | 8 | 2093 | |||
| XM_011535958.2 | XP_011534260.1 | 8 | 2081 | |||
| XM_011535959.2 | XP_011534261.1 | 7 | 1860 | |||
| XM_011535960.2 | XP_011534262.1 | 9 | 1844 | |||
| XM_011535961.2 | XP_011534263.1 | 6 | 1633 | |||
| XM_011535962.2 | XP_011534264.1 | 8 | 1561 | |||
| GenBank mRNA | AL832290.1 | - | 3 | 724 | ||
| BC039906.1 | - | 8 | 1905 | 6066481 | ||

PDSS2
Genomic Map