siRNA Details
| Gene Symbol: LPAR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142961.1 | NP_001136433.1 | 2 | 2584 | ||
| NM_020400.5 | NP_065133.1 | 2 | 2744 | |||
| GenBank mRNA | BC072394.1 | - | 2 | 2734 | 5751923 | |
More Information
Set Membership: |
|

LPAR5
Genomic Map