siRNA Details
| Gene Symbol: CD248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020404.2 | NP_065137.1 | 1 | 1517 | ||
| GenBank mRNA | AF279142.1 | - | 1 | 1505 | ||
| AJ295846.1 | - | 1 | 1517 | |||
| AK027290.1 | - | 1 | 639 | |||
| BC051340.1 | - | 1 | 1504 | 6339237 | ||
| BC104484.1 | - | 1 | 1517 | 40007855 | ||
| BC105633.1 | - | 1 | 1517 | 40007854 | ||
| KU215671.1 | - | 1 | 1455 | |||
| KU257670.1 | - | 1 | 1449 | |||

CD248
Genomic Map