siRNA Details
| Gene Symbol: DOLPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135917.1 | NP_001129389.1 | 7 | 1912 | ||
| NM_020438.4 | NP_065171.2 | 8 | 2041 | |||
| NR_072983.1 | 6 | 1827 | ||||
| GenBank mRNA | BC009493.2 | - | 8 | 2011 | ||
| BC033686.1 | - | 8 | 2019 | 5195127 | ||
| BC040152.1 | - | 6 | 1797 | |||

DOLPP1
Genomic Map