siRNA Details
| Gene Symbol: SPRYD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020456.3 | NP_065189.1 | 2 | 444 | ||
| XM_017020677.1 | XP_016876166.1 | 2 | 444 | |||
| GenBank mRNA | AF052154.1 | - | 2 | 243 | ||
| AF055016.1 | - | 2 | 243 | |||
| AF334405.1 | - | 2 | 283 | |||
| AK308818.1 | - | 2 | 277 | |||
| AK316576.1 | - | 2 | 250 | |||
| DA485722.1 | - | 2 | 445 | |||
| DN992366.1 | - | 2 | 440 | |||

SPRYD7
Genomic Map