siRNA Details
| Gene Symbol: THAP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020457.2 | NP_065190.2 | 1 | 982 | ||
| GenBank mRNA | AB015338.1 | - | 1 | 59 | ||
| AK289367.1 | - | 1 | 958 | |||
| BC012182.1 | - | 1 | 941 | 4554554 | ||
| EU869208.1 | - | 1 | 351 | |||
| L10378.1 | - | 1 | 819 | |||
| L10379.1 | - | 1 | 819 | |||

THAP11
Genomic Map