siRNA Details
| Gene Symbol: THAP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020457.2 | NP_065190.2 | 1 | 1544 | ||
| GenBank mRNA | BC012182.1 | - | 1 | 1503 | 4554554 | |
| L10378.1 | - | 1 | 1381 | |||
| L10379.1 | - | 1 | 1381 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

THAP11| Gene Symbol: THAP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020457.2 | NP_065190.2 | 1 | 1544 | ||
| GenBank mRNA | BC012182.1 | - | 1 | 1503 | 4554554 | |
| L10378.1 | - | 1 | 1381 | |||
| L10379.1 | - | 1 | 1381 | |||