siRNA Details
| Gene Symbol: C11orf16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020643.2 | NP_065694.2 | 4 | 571 | ||
| XM_017018013.1 | XP_016873502.1 | 3 | 1032 | |||
| GenBank mRNA | AK074455.1 | - | 4 | 666 | ||
| AK223377.1 | - | 4 | 666 | |||
| BC027865.1 | - | 4 | 553 | 5203029 | ||

C11orf16
Genomic Map