siRNA Details
| Gene Symbol: ZNF287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020653.2 | NP_065704.2 | 2 | 699 | ||
| XM_011523968.2 | XP_011522270.1 | 2 | 1145 | |||
| XM_017024887.1 | XP_016880376.1 | 2 | 906 | |||
| GenBank mRNA | AF217227.1 | - | 2 | 643 | ||
| BC032002.1 | - | 1 | 319 | |||
| BC041159.1 | - | 2 | 864 | |||
| BC057791.1 | - | 2 | 699 | |||
| BC101487.1 | - | 1 | 318 | 8068993 | ||
| BC113363.1 | - | 1 | 322 | 8322415 | ||
| CR457194.1 | - | 1 | 225 | |||

ZNF287
Genomic Map