siRNA Details
| Gene Symbol: SCAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145444.1 | NP_001138916.1 | 6 | 1083 | ||
| NM_001145445.1 | NP_001138917.1 | 7 | 1128 | |||
| NM_020706.2 | NP_065757.1 | 7 | 1128 | |||
| XM_006724035.2 | XP_006724098.1 | 7 | 1109 | |||
| XM_006724036.2 | XP_006724099.1 | 7 | 1109 | |||
| XM_017028415.1 | XP_016883904.1 | 7 | 1089 | |||
| XM_017028416.1 | XP_016883905.1 | 7 | 974 | |||
| GenBank mRNA | AB032998.1 | - | 1 | 116 | ||
| AF023142.1 | - | 7 | 732 | |||
| AK025748.1 | - | 2 | 147 | |||
| AK057840.1 | - | 7 | 971 | |||
| AK308406.1 | - | 6 | 934 | |||
| AL117417.1 | - | 2 | 150 | |||
| AL834304.1 | - | 7 | 908 | |||
| BC014921.1 | - | 7 | 1105 | 3907610 | ||
| BC043353.1 | - | 7 | 1085 | 6019414 | ||
| BC052286.1 | - | 7 | 959 | 5442227 | ||
| BC064990.1 | - | 7 | 959 | 5432277 | ||

SCAF4
Genomic Map