siRNA Details
| Gene Symbol: NLGN4X | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282145.1 | NP_001269074.1 | 7 | 6036 | ||
| NM_001282146.1 | NP_001269075.1 | 6 | 5711 | |||
| NM_020742.3 | NP_065793.1 | 6 | 5546 | |||
| NM_181332.2 | NP_851849.1 | 6 | 5729 | |||
| XM_005274564.2 | XP_005274621.1 | 7 | 5783 | |||
| XM_005274565.2 | XP_005274622.1 | 7 | 5766 | |||
| XM_005274566.4 | XP_005274623.1 | 7 | 5710 | |||
| XM_006724504.3 | XP_006724567.1 | 7 | 5651 | |||
| XM_011545547.2 | XP_011543849.1 | 7 | 5785 | |||
| XM_011545548.2 | XP_011543850.1 | 7 | 5949 | |||
| XM_017029690.1 | XP_016885179.1 | 7 | 5595 | |||
| XM_017029691.1 | XP_016885180.1 | 6 | 5725 | |||
| XM_017029692.1 | XP_016885181.1 | 6 | 5650 | |||
| XM_017029693.1 | XP_016885182.1 | 6 | 5535 | |||

NLGN4X
Genomic Map