siRNA Details
| Gene Symbol: KLHL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001292003.1 | NP_001278932.1 | 3 | 807 | ||
| NM_020803.4 | NP_065854.3 | 3 | 764 | |||
| GenBank mRNA | AB037799.2 | - | 2 | 447 | ||
| AK128827.1 | - | 3 | 807 | |||
| BC041384.2 | - | 3 | 714 | 5275946 | ||
| BX538173.1 | - | 3 | 681 | |||
| BX640727.1 | - | 3 | 674 | |||
| BX647454.1 | - | 3 | 699 | |||

KLHL8
Genomic Map