siRNA Details
| Gene Symbol: ZNF471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321768.1 | NP_001308697.1 | 4 | 2465 | ||
| NM_020813.3 | NP_065864.2 | 5 | 2592 | |||
| XM_011527148.1 | XP_011525450.1 | 5 | 5419 | |||
| XM_017027032.1 | XP_016882521.1 | 4 | 5292 | |||
| GenBank mRNA | AB037817.1 | - | 2 | 2234 | ||
| AL831845.1 | - | 5 | 2592 | |||
| BC125221.1 | - | 5 | 2549 | 40124078 | ||
| BC125222.1 | - | 5 | 2550 | 40124080 | ||

ZNF471
Genomic Map