siRNA Details
Gene Symbol: PITPNM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300801.1 | NP_001287730.1 | 5 | 661 | ||
NM_020845.2 | NP_065896.1 | 5 | 600 | |||
XM_011538590.2 | XP_011536892.1 | 5 | 625 | |||
GenBank mRNA | AB040890.2 | - | 6 | 682 | ||
AF334585.1 | - | 4 | 539 | |||
BC141813.1 | - | 5 | 701 |
