siRNA Details
| Gene Symbol: KLHL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020866.2 | NP_065917.1 | 2 | 1429 | ||
| XM_017020678.1 | XP_016876167.1 | 2 | 349 | |||
| XM_017020679.1 | XP_016876168.1 | 1 | 42 | |||
| GenBank mRNA | AB040923.1 | - | 2 | 1395 | ||
| AF252283.1 | - | 2 | 1263 | |||
| AK054609.1 | - | 2 | 641 | |||
| AK291435.1 | - | 2 | 1429 | |||
| BC022460.1 | - | 2 | 673 | 4819077 | ||

KLHL1
Genomic Map