siRNA Details
| Gene Symbol: KLHL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286725.1 | NP_001273654.1 | 5, 6 | 2036 | ||
| NM_020866.2 | NP_065917.1 | 6, 7 | 2219 | |||
| XM_017020678.1 | XP_016876167.1 | 6, 7 | 1139 | |||
| XM_017020679.1 | XP_016876168.1 | 6, 7 | 1726 | |||
| GenBank mRNA | AB040923.1 | - | 6, 7 | 2185 | ||
| AF252283.1 | - | 6, 7 | 2053 | |||
| AK054609.1 | - | 6, 7 | 1431 | |||
| AK291435.1 | - | 6, 7 | 2219 | |||
| AK295939.1 | - | 5, 6 | 1227 | |||
| BC022460.1 | - | 6, 7 | 1463 | 4819077 | ||

KLHL1
Genomic Map