siRNA Details
| Gene Symbol: LRRN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324188.1 | NP_001311117.1 | 3 | 1237 | ||
| NM_001324189.1 | NP_001311118.1 | 3 | 1160 | |||
| NM_020873.6 | NP_065924.3 | 2 | 955 | |||
| GenBank mRNA | AB040930.1 | - | 2 | 3454 | ||
| AB074161.1 | - | 1 | 188 | |||
| AK291717.1 | - | 2 | 539 | |||
| AY358312.1 | - | 1 | 302 | |||
| BC034947.2 | - | 2 | 518 | 4826337 | ||

LRRN1
Genomic Map